Learn More
Bioworld Tris/Glycine/SDS Buffer for Resolve Gel (1X), 20 PK(s)
Catalog No. 50198494
Encompass_Preferred
1X Tris/Glycine/SDS Buffer for Resolve GelBuffer System: Pre-weighed Powder mix in pouches(packs)1 pouch(pack)= 500 mL
Catalog No. 50-198-494 Supplier Bioworld Supplier No. 208400511
1X Tris/Glycine/SDS Buffer for Resolve GelBuffer System: Pre-weighed Powder mix in pouches(packs)1 pouch(pack)= 500 mL