Learn More
Accuflow Systems Saint-Gobain Sani-Tech Concentric Reducer, PVDF, Mini x 1 Ladish
Catalog No. NC3440542
Encompass_Preferred
The concentric reducer is a sanitary Tri-Clamp reducing connector designed for hygienic fluid transfer in bioprocessing and laboratory systems. The PVDF construction provides chemical resistance and a crevice-free inner bore for cleanability.
  • Concentric reducer for sanitary Tri-Clamp connections
  • Material: PVDF (Kynar), high chemical resistance
  • Configuration: Mini to 1 Ladish (mini x 1 ladish)
  • Typical dimensions: A = 0.750 in, B = 1.106 in, C = 3.000 in
  • Sanitary inner bore, crevice-free design, surface finish 5 Ra or better
  • Applications: bioprocessing, pharmaceutical, and laboratory fluid transfer
Catalog No. NC3440542 Supplier Accuflow Systems Supplier No. KAD100075L
The concentric reducer is a sanitary Tri-Clamp reducing connector designed for hygienic fluid transfer in bioprocessing and laboratory systems. The PVDF construction provides chemical resistance and a crevice-free inner bore for cleanability.
  • Concentric reducer for sanitary Tri-Clamp connections
  • Material: PVDF (Kynar), high chemical resistance
  • Configuration: Mini to 1 Ladish (mini x 1 ladish)
  • Typical dimensions: A = 0.750 in, B = 1.106 in, C = 3.000 in
  • Sanitary inner bore, crevice-free design, surface finish 5 Ra or better
  • Applications: bioprocessing, pharmaceutical, and laboratory fluid transfer