Learn More
Accuflow Systems Saint-Gobain Sani-Tech Silicone Hose, Braided, 3/8 in ID, 50 ft
Catalog No. NC1695179
Encompass_Preferred
Sani-Tech STHT-R braid-reinforced, platinum-cured silicone hose provides high-purity, flexible fluid transfer for higher-pressure bioprocess and laboratory applications. The reinforced construction increases working pressure while maintaining chemical inertness and wide temperature tolerance, making it suitable for pump lines, vessel transfers, and single-use assemblies.
  • Platinum-cured silicone construction for chemical resistance and purity.
  • Braid-reinforced for increased working pressure and dimensional stability.
  • Inner diameter 0.375 in (9.53 mm), outer diameter 0.625 in (15.88 mm).
  • Sold in 50 ft lengths.
  • Temperature range -80°F (-62°C) to 500°F (260°C).
  • Intended for pump and vessel transfer, filtration, and single-use assemblies.
Catalog No. NC1695179 Supplier Accuflow Systems Supplier No. STHTR0375F
Sani-Tech STHT-R braid-reinforced, platinum-cured silicone hose provides high-purity, flexible fluid transfer for higher-pressure bioprocess and laboratory applications. The reinforced construction increases working pressure while maintaining chemical inertness and wide temperature tolerance, making it suitable for pump lines, vessel transfers, and single-use assemblies.
  • Platinum-cured silicone construction for chemical resistance and purity.
  • Braid-reinforced for increased working pressure and dimensional stability.
  • Inner diameter 0.375 in (9.53 mm), outer diameter 0.625 in (15.88 mm).
  • Sold in 50 ft lengths.
  • Temperature range -80°F (-62°C) to 500°F (260°C).
  • Intended for pump and vessel transfer, filtration, and single-use assemblies.