Learn More
Alkali Scientific 1x8 StripFilm™, 8 Strips per Sheet, Sterile
SDP
Catalog No. 502388898
Encompass_Preferred
Designed for use with either 1x8 StripFilm™ ELISA plates or standard 96-well plates where rows must be selectively protected or accessed. Ideal for COVID-19 testing to seal a row of 8 wells after swabs are deposited into a deep-well plate. Certified DNase- & RNase- free. Designed to be used with both polystyrene and polypropylene plates. Precision cut to a smaller dimension to seal a single 8-well row of a 96-well plate or an ELISA strip-well plate. Each sheet contains 8 Mini Strips on a continuous backing liner for ease of use. Recommended for temperature range –40°C to +120°C. MiniStrips can be peeled and applied individually or two-at-a-time for increased efficiency.
Catalog No. 50-238-8898 Supplier Alkali Scientific Supplier No. SF8S
Designed for use with either 1x8 StripFilm™ ELISA plates or standard 96-well plates where rows must be selectively protected or accessed. Ideal for COVID-19 testing to seal a row of 8 wells after swabs are deposited into a deep-well plate. Certified DNase- & RNase- free. Designed to be used with both polystyrene and polypropylene plates. Precision cut to a smaller dimension to seal a single 8-well row of a 96-well plate or an ELISA strip-well plate. Each sheet contains 8 Mini Strips on a continuous backing liner for ease of use. Recommended for temperature range –40°C to +120°C. MiniStrips can be peeled and applied individually or two-at-a-time for increased efficiency.