Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Matrix Scientific (1Z)-N'-Hydroxyethanimidamide, 22059-22-9, MFCD00603514, 5g

Catalog No. 5001382326
Encompass_Preferred

Molecular Formula C2H6N2O, Purity 95+%, Molecular Weight 74.08

Catalog No. 50-013-82326 Supplier Matrix Scientific Supplier No. 0406925G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Molecular Formula C2H6N2O, Purity 95+%, Molecular Weight 74.08

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.