Learn More
Alkali Scientific 2.0mL Microtube with Cap, Clear, Sterile, with Self-Standing Bag; Stop-Pops™, 500/cs
SDP
Catalog No. 502335777
Encompass_Preferred
Qwik-Snap™ MicroTubes: Centrifugal Speed: 29,000 x g. Can withstand boiling up to 121℃ and freezing down to -80℃. Snaps into place for a secure seal. 100% polypropylene, RNase/DNase free. Clear, non-binding surface, graduated. Certified RNase, DNase and Pyrogen free. 500 sterile 2.0mL centrifuge tubes per pack.
Catalog No. 50-233-5777 Supplier Alkali Scientific Supplier No. C3002
Qwik-Snap™ MicroTubes: Centrifugal Speed: 29,000 x g. Can withstand boiling up to 121℃ and freezing down to -80℃. Snaps into place for a secure seal. 100% polypropylene, RNase/DNase free. Clear, non-binding surface, graduated. Certified RNase, DNase and Pyrogen free. 500 sterile 2.0mL centrifuge tubes per pack.