Learn More
USA Scientific Inc 2.0 mL con screw cap tube, org
Catalog No. 502745339
Encompass
2.0 mL conical base microcentrifuge tube, graduated, orange, with integrated TPE cap, 500/pack
Catalog No. 50-274-5339 Supplier USA Scientific Inc Supplier No. 14208303
2.0 mL conical base microcentrifuge tube, graduated, orange, with integrated TPE cap, 500/pack