Learn More
Chemglass Life Sciences Chemglass 2.0 mL Cryogenic Vial, Self-Standing, External Thread, Sterile
SDP
Catalog No. 502733392
Encompass_Preferred
A sterile 2.0 mL self-standing cryogenic vial designed for low-temperature sample storage and cryopreservation. Constructed from USP Class VI polypropylene and supplied sterile (E-beam), the vial is autoclavable and packaged for rack-based workflows in biobanking and laboratory cryostorage.
  • 2.0 mL capacity.
  • Self-standing, round-bottom, one-piece construction to maintain barcode adhesion at low temperatures.
  • External thread for secure cap closure.
  • Sterile; sterilized by E-beam.
  • Made from USP Class VI polypropylene; autoclavable.
  • Packaged in 9 x 9 racks (81 vials per rack), 12 racks per case (972 vials per case).
  • Suitable for long-term cryogenic storage and biobanking workflows.
Catalog No. 50-273-3392 Supplier Chemglass Life Sciences Supplier No. CGN4762003
A sterile 2.0 mL self-standing cryogenic vial designed for low-temperature sample storage and cryopreservation. Constructed from USP Class VI polypropylene and supplied sterile (E-beam), the vial is autoclavable and packaged for rack-based workflows in biobanking and laboratory cryostorage.
  • 2.0 mL capacity.
  • Self-standing, round-bottom, one-piece construction to maintain barcode adhesion at low temperatures.
  • External thread for secure cap closure.
  • Sterile; sterilized by E-beam.
  • Made from USP Class VI polypropylene; autoclavable.
  • Packaged in 9 x 9 racks (81 vials per rack), 12 racks per case (972 vials per case).
  • Suitable for long-term cryogenic storage and biobanking workflows.