Learn More
USA Scientific Inc 2.0 mL screw cap tube, amber
Catalog No. 502744872
Encompass
2.0 mL self-standing screw cap microcentrifuge tube, graduated, amber with amber cap, 500/pack
Catalog No. 50-274-4872 Supplier USA Scientific Inc Supplier No. 14209707
2.0 mL self-standing screw cap microcentrifuge tube, graduated, amber with amber cap, 500/pack