Learn More
Andwin Scientific 2.0 ML SELF-STANDING SAF-T-SEA
SDP
Catalog No. 50819773
Encompass_Preferred
The 0.5 mL self-standing screw cap tube is an ideal laboratory apparatus for centrifugation up to 20000 x g, vapor-phase liquid nitrogen storage, -80C freezers, boiling, and autoclaving. The tube's external threads prevent sample loss and seal failure. This product is autoclavable and free of RNase, DNase, DNA, and pyrogen. It comes in a pack of 500 tubes with natural-colored caps.
Catalog No. 50-819-773 Supplier Andwin Scientific Supplier No. 14209710
Add to Cart
Add to Cart
The 0.5 mL self-standing screw cap tube is an ideal laboratory apparatus for centrifugation up to 20000 x g, vapor-phase liquid nitrogen storage, -80C freezers, boiling, and autoclaving. The tube's external threads prevent sample loss and seal failure. This product is autoclavable and free of RNase, DNase, DNA, and pyrogen. It comes in a pack of 500 tubes with natural-colored caps.