Learn More
USA Scientific Inc 2.0 mL self-standing screw cap tube, natural, sterile
Catalog No. NC9692050
Encompass
2.0 mL self-standing screw cap microcentrifuge tube, graduated, natural with natural cap, sterile, 10 bags of 50 (500 tubes and caps)
Catalog No. NC9692050 Supplier USA Scientific Inc Supplier No. 14209710
2.0 mL self-standing screw cap microcentrifuge tube, graduated, natural with natural cap, sterile, 10 bags of 50 (500 tubes and caps)