Learn More
BioSpec Products 2.0 mm Zirconia Beads 1lb bottle
SDP
Catalog No. NC0419764
Encompass_Preferred
Zirconia beads have a density of 5.5g/cc (Twice as dense as glass beads - good for tough tissue).  Listed bead sizes are median (average) values. Because beads are sorted by sieves, bead size varies between +/-10% of a listed value. 1 lb bottle
Catalog No. NC0419764 Supplier BioSpec Products Supplier No. 11079124ZX
Zirconia beads have a density of 5.5g/cc (Twice as dense as glass beads - good for tough tissue).  Listed bead sizes are median (average) values. Because beads are sorted by sieves, bead size varies between +/-10% of a listed value. 1 lb bottle