Learn More
Sarstedt Inc 2.0 TUBE/SCR 500/CS
Supplier:  Sarstedt Inc 72.608
Encompass_Preferred
Micro tube 2 ml, polypropylene (PP), without cap. Conical base, recommended centrifugation 20000 x g. Diameter: 10,8 mm. Length of product: 44mm. Satisfies the IATA requirement.
Catalog No. NC0826136