Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glen Research 2.0 M Hexylammonium Acetate, HPLC grade, pH 7, volume 450 mL, ion-pairing buffer, for DMT-OFF oligonucleotide analysis and purification

Catalog No. NC2144973
Encompass_Preferred

This 2.0 M Hexylammonium Acetate solution is an HPLC grade buffer designed for ion-pairing reversed-phase HPLC (RP-HPLC) applications. It maintains a neutral pH of 7, ensuring compatibility with various analytical methods. With its strong interaction with reversed-phase resins, this concentrated buffer facilitates excellent separation based on oligonucleotide length, making it ideal for the analysis and purification of DMT-OFF oligonucleotides. The solution can be diluted twenty-fold to achieve a working concentration of 0.1 M, providing versatility in laboratory applications.

  • Concentration: 2.0 M
  • Grade: HPLC grade
  • pH: 7
  • Volume: 450 mL
  • Application type: Ion pairing, RP-HPLC buffer
  • Primary use: Analysis and purification of DMT-OFF oligonucleotides
  • Dilution factor: 1:20 (for 0.1 M working solution)
  • Interaction: Strong with reversed-phase resins
  • Separation principle: Oligonucleotide length

This item is not returnable. View return policy
Catalog No. NC2144973 Supplier Glen Research Supplier No. 60421057
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This 2.0 M Hexylammonium Acetate solution is an HPLC grade buffer designed for ion-pairing reversed-phase HPLC (RP-HPLC) applications. It maintains a neutral pH of 7, ensuring compatibility with various analytical methods. With its strong interaction with reversed-phase resins, this concentrated buffer facilitates excellent separation based on oligonucleotide length, making it ideal for the analysis and purification of DMT-OFF oligonucleotides. The solution can be diluted twenty-fold to achieve a working concentration of 0.1 M, providing versatility in laboratory applications.

  • Concentration: 2.0 M
  • Grade: HPLC grade
  • pH: 7
  • Volume: 450 mL
  • Application type: Ion pairing, RP-HPLC buffer
  • Primary use: Analysis and purification of DMT-OFF oligonucleotides
  • Dilution factor: 1:20 (for 0.1 M working solution)
  • Interaction: Strong with reversed-phase resins
  • Separation principle: Oligonucleotide length

This item is not returnable. View return policy
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.