Learn More
Analytical Sales & Services 2.0mL 96-Well Clear Flexi-Tier™ Block System with Amber Inserts
Catalog No. 5000806746
Encompass
Clear 96 Position Flexi-Tier Complete System. Includes: 2.0mL Amber Conical Glass Inserts, Base Plate, Pre-Scored PTFE/Silicone Flat Liner and Cover\n\nQty/Pkg: EACH
Catalog No. 50-008-06746 Supplier Analytical Sales & Services Supplier No. 96F220AC
Clear 96 Position Flexi-Tier Complete System. Includes: 2.0mL Amber Conical Glass Inserts, Base Plate, Pre-Scored PTFE/Silicone Flat Liner and Cover\n\nQty/Pkg: EACH