Learn More
Analytical Sales & Services 2.0mL 96-Well Clear Flexi-Tier™ Block System, Silanized
Catalog No. 5000806259
Encompass
Silanized 2.0mL 96-Well Clear Flexi-Tier™ Block System\n\nIncludes:\n• 2.0mL Silanized Conical Glass Inserts\n• Base Plate\n• 3 Spacers\n• Pre-Scored PTFE/Silicone Flat Liner\n• Cover\n\nQty/Pkg: Each
Catalog No. 50-008-06259 Supplier Analytical Sales & Services Supplier No. 96F220CS
Silanized 2.0mL 96-Well Clear Flexi-Tier™ Block System\n\nIncludes:\n• 2.0mL Silanized Conical Glass Inserts\n• Base Plate\n• 3 Spacers\n• Pre-Scored PTFE/Silicone Flat Liner\n• Cover\n\nQty/Pkg: Each