Learn More
Analytical Sales & Services 2.0mL 96-Well Opaque Flexi-Tier™ Block System
Catalog No. 5000806467
Encompass
2.0mL 96-Well Opaque Flexi-Tier Block System\n\nIncludes:\n• 2.0mL Conical Glass Inserts\n• Base Plate\n• 3 Spacers\n• Pre-Scored PTFE/Silicone Flat Liner\n• Cover\n\nQty/Pkg: EACH
Catalog No. 50-008-06467 Supplier Analytical Sales & Services Supplier No. 96F220
2.0mL 96-Well Opaque Flexi-Tier Block System\n\nIncludes:\n• 2.0mL Conical Glass Inserts\n• Base Plate\n• 3 Spacers\n• Pre-Scored PTFE/Silicone Flat Liner\n• Cover\n\nQty/Pkg: EACH