Learn More
Analytical Sales & Services 2.0mL Aluminum Block System, Solid Base with Conical Inserts
Catalog No. 5000806238
Encompass
2.0mL Aluminum Block System with Solid Base. Includes: Aluminum Base, Cover, Screws, Molded Silicone Liner and 2.0mL Clear Glass Conical Inserts Preassembled\n\nQty/Pkg: EACH
Catalog No. 50-008-06238 Supplier Analytical Sales & Services Supplier No. 96A120
2.0mL Aluminum Block System with Solid Base. Includes: Aluminum Base, Cover, Screws, Molded Silicone Liner and 2.0mL Clear Glass Conical Inserts Preassembled\n\nQty/Pkg: EACH