Learn More
Omega Biotek Inc 2 mL Collection Tubes, 5000/cs
Catalog No. NC1503375
Encompass
2.0 mL Tube, Capless, graduated. 100/pack, 50 pack/case.
Catalog No. NC1503375 Supplier Omega Biotek Inc Supplier No. AC137000
2.0 mL Tube, Capless, graduated. 100/pack, 50 pack/case.