Learn More
USA Scientific Inc 2.0 mL conical screw cap tube, blue cap
Catalog No. 507541465
Encompass_Preferred
2.0 mL conical base screw cap microcentrifuge tube, graduated, natural with blue cap, 500/pack
Catalog No. 50-754-1465 Supplier USA Scientific Inc Supplier No. 14208701
2.0 mL conical base screw cap microcentrifuge tube, graduated, natural with blue cap, 500/pack