Learn More
USA Scientific Inc 2.0 mL conical screw cap tube, violet cap
Catalog No. 507541469
Encompass_Preferred
2.0 mL conical base screw cap microcentrifuge tube, graduated, natural with violet cap, 500/pack
Catalog No. 50-754-1469 Supplier USA Scientific Inc Supplier No. 14208705
2.0 mL conical base screw cap microcentrifuge tube, graduated, natural with violet cap, 500/pack