Learn More
Chemglass Life Sciences Chemglass 2.0 mL Cryogenic Vial, Self-Standing, Internal Thread, Sterile (CGN-4766-011)
SDP
Catalog No. 502733342
Encompass_Preferred
The Chemglass 2.0 mL Cryogenic Vial is a sterile, self-standing vial designed for secure sample storage at ultra-low temperatures. It features an internal-thread closure and is manufactured from USP 6 grade raw materials for biocompatibility. Vials are supplied racked for organized handling and bulk storage.
  • 2.0 mL capacity.
  • Self-standing cryogenic vial.
  • Internal-thread closure for a secure seal.
  • Manufactured from USP 6 grade raw materials.
  • Sterile.
  • Packaged 9 x 9 per rack; 12 racks per case (972 vials per case).
Catalog No. 50-273-3342 Supplier Chemglass Life Sciences Supplier No. CGN4766011
The Chemglass 2.0 mL Cryogenic Vial is a sterile, self-standing vial designed for secure sample storage at ultra-low temperatures. It features an internal-thread closure and is manufactured from USP 6 grade raw materials for biocompatibility. Vials are supplied racked for organized handling and bulk storage.
  • 2.0 mL capacity.
  • Self-standing cryogenic vial.
  • Internal-thread closure for a secure seal.
  • Manufactured from USP 6 grade raw materials.
  • Sterile.
  • Packaged 9 x 9 per rack; 12 racks per case (972 vials per case).