Learn More
Alkali Scientific CellPro™ Externally Threaded Cryovials, 2.0mL, Self Standing, Sterile, 50/Bag, 500/pk
SDP
Catalog No. 502388932
Encompass_Preferred
CellPro™ Cryogenic Vials are manufactured from high visibility polypropylene for stability in extreme temperature conditions. CellPro™ Externally Threaded Cryogenic Vials provide a safe and secure tube for ultra-low temperature storage of valuable cell cultures, clinical hospital samples and biological samples. The silicone O-rings allow for expansion and contraction in liquid nitrogen environments associated with long term, -190°C storage.
Catalog No. 50-238-8932 Supplier Alkali Scientific Supplier No. VCE23
CellPro™ Cryogenic Vials are manufactured from high visibility polypropylene for stability in extreme temperature conditions. CellPro™ Externally Threaded Cryogenic Vials provide a safe and secure tube for ultra-low temperature storage of valuable cell cultures, clinical hospital samples and biological samples. The silicone O-rings allow for expansion and contraction in liquid nitrogen environments associated with long term, -190°C storage.