Learn More
Chemglass Life Sciences MicroCentrifuge Tube, 2.0mL, Clear, Round, Lock Cap
SDP
Catalog No. 501897713
Encompass_Preferred
• USP VI grade polypropylene material, free from heavy metal • RCF: 30,000 x g • Endotoxin level: <0.1EU • Clear graduations and frosted marking area • Special Safe-lock cap design provides secure lid sealing • Easy opening and closing • Temperature range: -80℃ to +121℃ • DNase/RNase-freeTubes are autoclavable at 121°C (250°F), under 15psi for 10–15 minutes with drying. Remove from autoclave immediately. Excessive exposure to autoclaving heat may cause warping or decreased structural integrity.
Catalog No. 50-189-7713 Supplier Chemglass Life Sciences Supplier No. CGN4310020
• USP VI grade polypropylene material, free from heavy metal • RCF: 30,000 x g • Endotoxin level: <0.1EU • Clear graduations and frosted marking area • Special Safe-lock cap design provides secure lid sealing • Easy opening and closing • Temperature range: -80℃ to +121℃ • DNase/RNase-freeTubes are autoclavable at 121°C (250°F), under 15psi for 10–15 minutes with drying. Remove from autoclave immediately. Excessive exposure to autoclaving heat may cause warping or decreased structural integrity.