Learn More
Biotang Inc 2.0 ml Microcentrifuge Tubes.26000g. 5000/CS .Superior clarity. Fit all standard rotors.Autoclavable. Dnase, Rnase, Human DNA, Pyrogen, PCR hibitors free.
Catalog No. 507515026 Shop All BIOTANG Inc Products
Encompass_Preferred
2.0 ml Microcentrifuge Tubes.26000g. 5000/CS .Superior clarity. Fit all standard rotors.Autoclavable. Dnase, Rnase, Human DNA, Pyrogen, PCR hibitors free.
Catalog No. 50-751-5026 Supplier Biotang Inc Supplier No. BT12030
2.0 ml Microcentrifuge Tubes.26000g. 5000/CS .Superior clarity. Fit all standard rotors.Autoclavable. Dnase, Rnase, Human DNA, Pyrogen, PCR hibitors free.