Learn More
Research Products International Corp Micro-Centrifuge Tubes, 2.0ml Capacity, Graduated 100 UL, Red, 500 per Case
SDP
Catalog No. 501537738
Encompass
Micro-Centrifuge Tubes have molded external graduation marks at 100µl increments that are clear and easy to read, similar in style to marks used on graduated glassware. Tubes have a flat frosted top and a frosted marking area on the side of tube for writing. Hinged lid provides a soft piercing point for inserting a needle. Made from a clear copolymer designed for high speed centrifugation and boiling.
Catalog No. 50-153-7738 Supplier Research Products International Corp Supplier No. 145520R
Micro-Centrifuge Tubes have molded external graduation marks at 100µl increments that are clear and easy to read, similar in style to marks used on graduated glassware. Tubes have a flat frosted top and a frosted marking area on the side of tube for writing. Hinged lid provides a soft piercing point for inserting a needle. Made from a clear copolymer designed for high speed centrifugation and boiling.