Learn More
USA Scientific Inc 2.0 mL low adhesion microcentrifuge tube, natural
Catalog No. NC9946978
Encompass
2.0 mL low adhesion polypropylene microcentrifuge tube, natural, graduated, 500/bag
Catalog No. NC9946978 Supplier USA Scientific Inc Supplier No. 14202600
2.0 mL low adhesion polypropylene microcentrifuge tube, natural, graduated, 500/bag