Learn More
NEST SCIENTIFIC INC 2.0ml Microcentrifuge Tube, Clear, Round, Lock Cap, 5000/cs
Catalog No. NC3429300
Encompass_Preferred
Nest microcentrifuge tubes are molded from USP grade VI polypropylene. Made using uses diamond polished molds for superior tolerances. Excellent sealing and low-retention perfomance without PFAS additives. Molded in an ISO 7 cleanroom and lot tested free of DNase, RNAase, DNA, cytoxins/endotoxins and BSE/TSE. cGMP compliant (21 CFR part 820/821).
Catalog No. NC3429300 Supplier NEST SCIENTIFIC INC Supplier No. 620011
Nest microcentrifuge tubes are molded from USP grade VI polypropylene. Made using uses diamond polished molds for superior tolerances. Excellent sealing and low-retention perfomance without PFAS additives. Molded in an ISO 7 cleanroom and lot tested free of DNase, RNAase, DNA, cytoxins/endotoxins and BSE/TSE. cGMP compliant (21 CFR part 820/821).