Learn More
Chemglass Life Sciences 2.0mL Micro Centrifuge Tube, Graduated, Self-Standing, Natural, CLIKLOK Flat Cap
SDP
Catalog No. 501214627
Encompass_Preferred
2.0mL Micro Centrifuge Tube, Graduated, Self-Standing, Natural, CLIKLOK Flat Cap, These 2.0mL graduated, rugged tubes are made of laboratory grade polypropylene suitable to withstand the stress of high speed centrifugation up to 20,000g. They offer a special locking clasp to produce a more secure closure. They are graduated at 0.5, 1.0 and 1.5mL. They are autoclavable up to 121C. They have also been designed with a pierceable lid. The tubes incorporate an etched surface on the side for sample identification. The tubes offer a Clicklok seal especially designed to reduce popping where freezing, boiling or shipping is required. Available in four colors. Packaged in tamperproof, re-sealable safety-lock bags.
Catalog No. 50-121-4627 Supplier Chemglass Life Sciences Supplier No. CLS430012N
2.0mL Micro Centrifuge Tube, Graduated, Self-Standing, Natural, CLIKLOK Flat Cap, These 2.0mL graduated, rugged tubes are made of laboratory grade polypropylene suitable to withstand the stress of high speed centrifugation up to 20,000g. They offer a special locking clasp to produce a more secure closure. They are graduated at 0.5, 1.0 and 1.5mL. They are autoclavable up to 121C. They have also been designed with a pierceable lid. The tubes incorporate an etched surface on the side for sample identification. The tubes offer a Clicklok seal especially designed to reduce popping where freezing, boiling or shipping is required. Available in four colors. Packaged in tamperproof, re-sealable safety-lock bags.