Learn More
Ace Glass, Inc. 2mL glass Pestle with 6mm rod
SDP
Catalog No. 50974566
Encompass_Preferred
2mL glass Pestle with 6mm rod, for use with 2mL tapered tube (50974565) as a tissue grinder. The rod can be used with a 5097694 Flex-Grip Chuck.
This item is not returnable. View return policy
Catalog No. 50-974-566 Supplier Ace Glass, Inc. Supplier No. 832506
2mL glass Pestle with 6mm rod, for use with 2mL tapered tube (50974565) as a tissue grinder. The rod can be used with a 5097694 Flex-Grip Chuck.