Learn More
Alkali Scientific 2.0mL Qwik-Snap™ Microcentrifuge Tubes, 500/Pk
SDP
Catalog No. 503026566
Encompass_Preferred
Alkali Scientific Microcentrifuge Tubes 2.0mL Qwik-Snap are made of premium polypropylene plastic which allows for a crystal clear tube and high centrifugal strength. They feature a secure-snap cap that ensures a strong seal and eliminates leakage and evaporation.  Qwik-Snap MicroTubes are also available in assorted colors. Capable of withstanding boiling temperatures up to 121° C and freezing down to -80°C, the Qwik-Snap micro centrifuge tubes are the perfect choice for your research. Centrifugal Speed: 29,000 x g Withstands boiling up to 121°C and freezing down to -80°C Snaps into place for a secure seal 100% polypropylene human DNA free Clear, non-binding surface, graduated Certified RNase, DNase & Pyrogen free
Catalog No. 50-302-6566 Supplier Alkali Scientific Supplier No. C3020
Add to Cart
Add to Cart
Alkali Scientific Microcentrifuge Tubes 2.0mL Qwik-Snap are made of premium polypropylene plastic which allows for a crystal clear tube and high centrifugal strength. They feature a secure-snap cap that ensures a strong seal and eliminates leakage and evaporation.  Qwik-Snap MicroTubes are also available in assorted colors. Capable of withstanding boiling temperatures up to 121° C and freezing down to -80°C, the Qwik-Snap micro centrifuge tubes are the perfect choice for your research. Centrifugal Speed: 29,000 x g Withstands boiling up to 121°C and freezing down to -80°C Snaps into place for a secure seal 100% polypropylene human DNA free Clear, non-binding surface, graduated Certified RNase, DNase & Pyrogen free