Learn More
Analytical Sales & Services 2.0mL Silanized Amber Glass Conical Vials with Marking Spots in Vial Loader
Catalog No. 5000807181
Encompass
Advantage 2.0mL Silanized Amber Glass Conical Vials with Marking Spots in Vial Loader\n\nQty/Pkg: EACH
Catalog No. 50-008-07181 Supplier Analytical Sales & Services Supplier No. 96FV20AS
Advantage 2.0mL Silanized Amber Glass Conical Vials with Marking Spots in Vial Loader\n\nQty/Pkg: EACH