Learn More
CELLTREAT Scientific Products 2.0mL Self-Standing Micro Centrifuge Tube, Non-sterile
SDP
Catalog No. 50202027
Encompass_Preferred
CELLTREAT Scientific Products 2.0mL Self-standing Micro Centrifuge Tube. Tubes have frosted marking spot and engraved graduations. Attached flat top cap is easy to open and close with one hand. Temperature range of -80¿C to 120¿C. Rnase/Dnase free and non-pyrogenic. Max force (RCF) g 20000. Every case is printed with the lot number for quality traceability. Packaged non-sterile, 500/re-sealable bag, 5000/case.
Catalog No. 50-202-027 Supplier CELLTREAT Scientific Products Supplier No. 229448
CELLTREAT Scientific Products 2.0mL Self-standing Micro Centrifuge Tube. Tubes have frosted marking spot and engraved graduations. Attached flat top cap is easy to open and close with one hand. Temperature range of -80¿C to 120¿C. Rnase/Dnase free and non-pyrogenic. Max force (RCF) g 20000. Every case is printed with the lot number for quality traceability. Packaged non-sterile, 500/re-sealable bag, 5000/case.