Learn More
USA Scientific Inc Seal-Rite 2.0 mL microcentrifuge tube, natural, case of 10 bags
Catalog No. NC0742040
Encompass_Preferred
Seal-Rite 2.0 mL graduated microcentrifuge tube, natural, polypropylene, 500/bag, case of 10 bags (5000 tubes)
Catalog No. NC0742040 Supplier USA Scientific Inc Supplier No. 16202720
Seal-Rite 2.0 mL graduated microcentrifuge tube, natural, polypropylene, 500/bag, case of 10 bags (5000 tubes)