Learn More
Ace Glass, Inc. 2mL all-glass tapered tube with a pouring lip
SDP
Catalog No. 50974565
Encompass_Preferred
2mL all-glass tapered tube with a pouring lip, for use with a pestle as a tissue grinder. The conical area of the tube is for initial grinding, and the cylindrical area is for final homogenization. Interchangeably ground to 0.1-0.15mm clearance. Capacity represents volume with pestle inserted.
This item is not returnable. View return policy
Catalog No. 50-974-565 Supplier Ace Glass, Inc. Supplier No. 832504
2mL all-glass tapered tube with a pouring lip, for use with a pestle as a tissue grinder. The conical area of the tube is for initial grinding, and the cylindrical area is for final homogenization. Interchangeably ground to 0.1-0.15mm clearance. Capacity represents volume with pestle inserted.