Learn More
USA Scientific Inc 2.0mL v-bot screw cap tube
Catalog No. 502745340
Encompass
2.0 mL conical base microcentrifuge tube, graduated, yellow, with integrated TPE cap, 500/pack
Catalog No. 50-274-5340 Supplier USA Scientific Inc Supplier No. 14208306
Add to Cart
Add to Cart
2.0 mL conical base microcentrifuge tube, graduated, yellow, with integrated TPE cap, 500/pack