Learn More
USA Scientific Inc 2.0mL v-bot screw cap tube
Catalog No. 502745271
Encompass
2.0 mL conical base microcentrifuge tube, graduated, red, with integrated TPE cap, 500/pack
Catalog No. 50-274-5271 Supplier USA Scientific Inc Supplier No. 14208304
2.0 mL conical base microcentrifuge tube, graduated, red, with integrated TPE cap, 500/pack