Learn More
USA Scientific Inc 2.0mL v-bot screw cap tube
Supplier:  USA Scientific Inc 14208300
Encompass
2.0 mL conical base screw cap microcentrifuge tube, graduated, natural, with integrated TPE natural color cap, 500/pack
Catalog No. 50-274-5548