Learn More
Chemglass Life Sciences 2.0mL Micro Centrifuge Tube, Graduated, Conical, Natural, CLIKLOK Flat Cap
SDP
Catalog No. 501214623
Encompass_Preferred
2.0mL Micro Centrifuge Tube, Graduated, Conical, Natural, CLIKLOK Flat Cap. These 2.0mL graduated, rugged tubes are made of laboratory grade polypropylene suitable to withstand the stress of high speed centrifugation up to 20,000g. They offer a special locking clasp to produce a more secure closure. They are graduated at 0.5, 1.0 and 1.5mL. They are autoclavable up to 121¿C. They have also been designed with a pierceable lid. The tubes incorporate an etched surface on the side for sample identification. The tubes offer a Clicklok seal especially designed to reduce popping where freezing, boiling or shipping is required. Available in four colors. Packaged in tamperproof, re-sealable safety-lock bags.This superior quality Clicklok system works in two stages:1. If only spinning is to be done, a slight push of the cap over the tube is required.2. For a complete seal, press further on snap cap until a ¿Click¿ is heard.
Catalog No. 50-121-4623 Supplier Chemglass Life Sciences Supplier No. CLS430002N
2.0mL Micro Centrifuge Tube, Graduated, Conical, Natural, CLIKLOK Flat Cap. These 2.0mL graduated, rugged tubes are made of laboratory grade polypropylene suitable to withstand the stress of high speed centrifugation up to 20,000g. They offer a special locking clasp to produce a more secure closure. They are graduated at 0.5, 1.0 and 1.5mL. They are autoclavable up to 121¿C. They have also been designed with a pierceable lid. The tubes incorporate an etched surface on the side for sample identification. The tubes offer a Clicklok seal especially designed to reduce popping where freezing, boiling or shipping is required. Available in four colors. Packaged in tamperproof, re-sealable safety-lock bags.This superior quality Clicklok system works in two stages:1. If only spinning is to be done, a slight push of the cap over the tube is required.2. For a complete seal, press further on snap cap until a ¿Click¿ is heard.