Learn More
Med Vet International 2.0mm Cortical Self-Tapping Screws - 2.0mm Cortical Self-Tapping Screw 8mm, J0416BS, Each
Catalog No. 5000582672
Encompass_Preferred
Available in 6mm-24mm lengths, the spherical head of the screw produces compression when used in conjunction with 2mm compression plate. Self-tapping threads sold in single packs. Self-Tapping screws save time and work extremely well in most circumstances especally in the proximal tibia for TPLO and TTO. Cortical Screw. Self-Tapping Threads. 8mm L
Catalog No. 50-005-82672 Supplier Med Vet International Supplier No. JOR416BS
Available in 6mm-24mm lengths, the spherical head of the screw produces compression when used in conjunction with 2mm compression plate. Self-tapping threads sold in single packs. Self-Tapping screws save time and work extremely well in most circumstances especally in the proximal tibia for TPLO and TTO. Cortical Screw. Self-Tapping Threads. 8mm L