Learn More
Med Vet International 2.0mm Drill Bit Hard, J0372BH
Catalog No. 5000582937
Encompass_Preferred
TTA (Tibial Tuberosity Advancement). TTA is preferred by some surgeons. Further details on the actual surgery can be seen on www.JorVet.com or the instructional DVD provided. This system is available in either titanium or stainless steel; titanium is more expensive but offers better bio-compatibility and fatigue strength.TTA Instrumentation - 2.0mm Drill Bit Hard
Catalog No. 50-005-82937 Supplier Med Vet International Supplier No. JOR372BH
TTA (Tibial Tuberosity Advancement). TTA is preferred by some surgeons. Further details on the actual surgery can be seen on www.JorVet.com or the instructional DVD provided. This system is available in either titanium or stainless steel; titanium is more expensive but offers better bio-compatibility and fatigue strength.TTA Instrumentation - 2.0mm Drill Bit Hard