Learn More
IBI SCIENTIFIC 3P 2.0MM X 16 TOOTH COMBS, 2/PK
Catalog No. NC0718291
Encompass_Preferred
2.0mm x 16 tooth Comb, 2/PK. Compatible with the IB53000 MP-1015 (Multi-Purpose) Horizontal Electrophoresis System (10x15cm). Well width = 3.5mm. Sample volume/mm gel thickness = 7.0ul. Manufactured from Delrin to provide years of service.
Catalog No. NC0718291 Supplier IBI SCIENTIFIC 3P Supplier No. IB52100
2.0mm x 16 tooth Comb, 2/PK. Compatible with the IB53000 MP-1015 (Multi-Purpose) Horizontal Electrophoresis System (10x15cm). Well width = 3.5mm. Sample volume/mm gel thickness = 7.0ul. Manufactured from Delrin to provide years of service.