Learn More
IBI SCIENTIFIC 3P 2.0MM X20 TOOTH COMBS, 2/PK
Catalog No. NC0134242
Encompass_Preferred
2.0mm x 20 tooth Comb, 2/PK. Compatible with the IB53000 MP-1015 (Multi-Purpose) Horizontal Electrophoresis System (10x15cm). Well width = 2.6mm. Sample volume/mm gel thickness = 5.2ul. Manufactured from Delrin to provide years of service.
Catalog No. NC0134242 Supplier IBI SCIENTIFIC 3P Supplier No. IB52170
2.0mm x 20 tooth Comb, 2/PK. Compatible with the IB53000 MP-1015 (Multi-Purpose) Horizontal Electrophoresis System (10x15cm). Well width = 2.6mm. Sample volume/mm gel thickness = 5.2ul. Manufactured from Delrin to provide years of service.