Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HF Scientific Inc Reference Standard Solution | Chlorine Photometer | 2.0ppm | 118ml

Catalog No. NC1410310
Encompass_Preferred

Reference Standard Solution | Chlorine Photometer | 2.0ppm | 118ml

Catalog No. NC1410310 Supplier HF Scientific Inc Supplier No. 10451A
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Reference Standard Solution | Chlorine Photometer | 2.0ppm | 118ml

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.