Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-(1-(2-amino-2-oxoethyl)cyclohexyl)acetic acid | 99189-60-3 | MFCD02181086 | 97.0% | 199.25 | C10H17NO3 | 50 G
SDP

Catalog No. 5000301625
Encompass_Preferred

2-(1-(2-amino-2-oxoethyl)cyclohexyl)acetic acid (CAS 99189-60-3) is a small-molecule biochemical reagent supplied as a white to off-white solid for research use. It is used in biochemical assays and studies of protein and enzyme function, and is available in laboratory-scale quantities.

  • White to off-white solid powder.
  • Purity 97.0%.
  • Molecular formula C10H17NO3; molecular weight 199.25.
  • Available in multiple sizes for lab-scale experiments.
  • Supplied with data sheet and COA for quality control.

Catalog No. 50-003-01625 Supplier Medchemexpress LLC Supplier No. HYW02583050G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

2-(1-(2-amino-2-oxoethyl)cyclohexyl)acetic acid (CAS 99189-60-3) is a small-molecule biochemical reagent supplied as a white to off-white solid for research use. It is used in biochemical assays and studies of protein and enzyme function, and is available in laboratory-scale quantities.

  • White to off-white solid powder.
  • Purity 97.0%.
  • Molecular formula C10H17NO3; molecular weight 199.25.
  • Available in multiple sizes for lab-scale experiments.
  • Supplied with data sheet and COA for quality control.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.