Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-(1-(2-amino-2-oxoethyl)cyclohexyl)acetic acid | 99189-60-3 | MFCD02181086 | 97.0% | 199.25 g/mol | C10H17NO3 | 100 G
SDP

Catalog No. 5000301642
Encompass_Preferred

2-(1-(2-Amino-2-oxoethyl)cyclohexyl)acetic acid is a small-molecule biochemical reagent supplied as a white to off-white solid with a reported purity of 97.0%. It is intended for research use; follow supplier guidance for handling, storage, and safety.

  • Molecular formula: C10H17NO3.
  • Molecular weight: 199.25 g/mol.
  • Purity: 97.0%.
  • Appearance: white to off-white solid.
  • Recommended storage: powder at -20°C (3 years) or 4°C (2 years); in solvent at -80°C (6 months).

Catalog No. 50-003-01642 Supplier Medchemexpress LLC Supplier No. HYW025830100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-(1-(2-Amino-2-oxoethyl)cyclohexyl)acetic acid is a small-molecule biochemical reagent supplied as a white to off-white solid with a reported purity of 97.0%. It is intended for research use; follow supplier guidance for handling, storage, and safety.

  • Molecular formula: C10H17NO3.
  • Molecular weight: 199.25 g/mol.
  • Purity: 97.0%.
  • Appearance: white to off-white solid.
  • Recommended storage: powder at -20°C (3 years) or 4°C (2 years); in solvent at -80°C (6 months).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.