Learn More
US Plastic Corporation 2-1/2 Gallon Nalgene Wide Mouth LDPE Carboy Modified by Tamco with a Fast Draw Off Spigot
SDP
Catalog No. NC1456866
Encompass
Our finest polyethylene carboy, featuring a spigot that has 3/4"-16 UNF with a 1" long thread, flow rate of 1.18 gpm. This carboy is lab quality, heavy duty, leakproof, and resistant to most chemicals.
Catalog No. NC1456866 Supplier US Plastic Corporation Supplier No. 65015
Our finest polyethylene carboy, featuring a spigot that has 3/4"-16 UNF with a 1" long thread, flow rate of 1.18 gpm. This carboy is lab quality, heavy duty, leakproof, and resistant to most chemicals.