Learn More
Cole-Parmer Cole-Parmer Egg-Shaped Stir Bar, 2-1/2" L x 3/4" Dia; 1/Pk
Catalog No. NC9408776 Shop All Cole-Parmer Products
Encompass_Preferred
Egg-shaped stir bars have oval magnets—ideal for use in round-bottom flasks. Shape conforms to flask bottom and ensures complete mixing.
Catalog No. NC9408776 Supplier Cole-Parmer Supplier No. 0476875
Egg-shaped stir bars have oval magnets—ideal for use in round-bottom flasks. Shape conforms to flask bottom and ensures complete mixing.