Learn More
Medchemexpress LLC 2-[1-[(2S,3S)-2-(2,3-dihydro-1H-inden-2-ylmethyl)-3-(3,5-dimethoxy-4-methylphenyl)...]acetic acid | 856691-44-6 | 99.9% | 521.60 | C30H35NO7 | 25 MG
SDP
Catalog No. 5000227749
Encompass_Preferred
ONO-9780307 is a synthetic antagonist of the lysophosphatidic acid receptor 1 (LPA1) with high potency (IC50 = 2.7 nM). It has been used as a pharmacological tool and as a co-crystallized ligand for human LPA1 (PDB 4Z34). Supplied as a 25 mg solid, the compound has formula C30H35NO7 and a molecular weight of 521.60; store protected from light and follow recommended solvent storage conditions.
  • High potency LPA1 antagonist (IC50 2.7 nM).
  • Validated for receptor binding and structural biology applications.
  • High purity confirmed by certificate of analysis (>99.9%).
  • Supplied as a solid suitable for dissolution in common solvents (e.g., DMSO).
  • Short- and long-term storage guidance provided for solid and solvent solutions.
Catalog No. 50-002-27749 Supplier Medchemexpress LLC Supplier No. HY11744425MG
ONO-9780307 is a synthetic antagonist of the lysophosphatidic acid receptor 1 (LPA1) with high potency (IC50 = 2.7 nM). It has been used as a pharmacological tool and as a co-crystallized ligand for human LPA1 (PDB 4Z34). Supplied as a 25 mg solid, the compound has formula C30H35NO7 and a molecular weight of 521.60; store protected from light and follow recommended solvent storage conditions.
  • High potency LPA1 antagonist (IC50 2.7 nM).
  • Validated for receptor binding and structural biology applications.
  • High purity confirmed by certificate of analysis (>99.9%).
  • Supplied as a solid suitable for dissolution in common solvents (e.g., DMSO).
  • Short- and long-term storage guidance provided for solid and solvent solutions.