Learn More
Medchemexpress LLC 2,1,3-benzothiadiazole | 273-13-2 | MFCD00005809 | 98.0% | 136.18 g/mol | C6H4N2S | 25 G
SDP
Catalog No. 5000302680
Encompass_Preferred
2,1,3-Benzothiadiazole is a bicyclic heterocyclic organic compound (C6H4N2S) used as a biochemical reagent and synthetic building block in life-science and materials research. It finds application in organic synthesis, agrochemical studies, and optoelectronic materials due to its thiadiazole ring, conjugated aromatic system, and thermal stability.
  • Bicyclic heterocycle with an electron-withdrawing thiadiazole ring.
  • Useful building block for organic synthesis and derivatization.
  • Applicable in agrochemical and optoelectronic materials research.
  • Available in high-purity grades for research use (commonly ~98%).
  • Solid at ambient conditions and stable under typical laboratory storage.
Catalog No. 50-003-02680 Supplier Medchemexpress LLC Supplier No. 25GHYW017038
2,1,3-Benzothiadiazole is a bicyclic heterocyclic organic compound (C6H4N2S) used as a biochemical reagent and synthetic building block in life-science and materials research. It finds application in organic synthesis, agrochemical studies, and optoelectronic materials due to its thiadiazole ring, conjugated aromatic system, and thermal stability.
  • Bicyclic heterocycle with an electron-withdrawing thiadiazole ring.
  • Useful building block for organic synthesis and derivatization.
  • Applicable in agrochemical and optoelectronic materials research.
  • Available in high-purity grades for research use (commonly ~98%).
  • Solid at ambient conditions and stable under typical laboratory storage.